N-(6-Chloro-2-pyridinyl)-N-(2-furylmethyl)amine
Supplier: Matrix Scientific
053184-500MG
053184-2.5G
101902-624EA
364.62
USD
101902-624
101902-626
N-(6-Chloro-2-pyridinyl)-N-(2-furylmethyl)amine
N-(6-Chloro-2-pyridinyl)-N-(2-furylmethyl)amine
Hazard: Irritant
Formula:
C₁₀H₉ClN₂O |
MDL Number:
MFCD13562618 CAS Number: 1220035-17-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...