N-(5-Bromo-2-pyridinyl)-N-(2-furylmethyl)amine

Supplier: Matrix Scientific

053183-500MG 053183-2.5G
101902-620EA 364.62 USD
101902-620 101902-622
N-(5-Bromo-2-pyridinyl)-N-(2-furylmethyl)amine
N-(5-Bromo-2-pyridinyl)-N-(2-furylmethyl)amine

Hazard: Irritant

Formula: C₁₀H₉BrN₂O
MDL Number: MFCD11116385
CAS Number: 1036508-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR