2-[(2-Furylmethyl)amino]nicotinic acid
Supplier: Matrix Scientific
053179-500MG
053179-2.5G
101902-604EA
384.62
USD
101902-604
101902-606
2-[(2-Furylmethyl)amino]nicotinic acid
2-[(2-Furylmethyl)amino]nicotinic acid
Hazard: Irritant
Formula:
C₁₁H₁₀N₂O₃ |
MDL Number:
MFCD11106739 CAS Number: 1019127-14-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...