N-(6-Chloro-2-pyrazinyl)-N-(2-furylmethyl)amine

Supplier: Matrix Scientific

053177-500MG 053177-2.5G
101902-596EA 364.62 USD
101902-596 101902-598
N-(6-Chloro-2-pyrazinyl)-N-(2-furylmethyl)amine
N-(6-Chloro-2-pyrazinyl)-N-(2-furylmethyl)amine

Hazard: Irritant

Formula: C₉H₈ClN₃O
MDL Number: MFCD13562616
CAS Number: 629658-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR