N-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-N-(2-furylmethyl)amine
Supplier: Matrix Scientific
053176-500MG
053176-2.5G
101902-592EA
364.62
USD
101902-592
101902-594
N-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-N-(2-furylmethyl)amine
N-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-N-(2-furylmethyl)amine
Hazard: Irritant
Formula:
C₁₁H₈ClF₃N₂O |
MDL Number:
MFCD11118706 CAS Number: 1036578-95-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...