N-(6-Chloro-4-pyrimidinyl)-N-(2-furylmethyl)amine
Supplier: Matrix Scientific
Synonyms:
6-Chloro-4-(furfurylamino)pyrimidine
053175-500MG
053175-2.5G
101902-588EA
166.15
USD
101902-588
101902-590
N-(6-Chloro-4-pyrimidinyl)-N-(2-furylmethyl)amine
N-(6-Chloro-4-pyrimidinyl)-N-(2-furylmethyl)amine
Hazard: Irritant
Formula:
C₉H₈ClN₃O MW: 209.633 g/mol |
MDL Number:
MFCD11655644 CAS Number: 99846-86-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...