N-(5-Bromo-4-methyl-2-pyridinyl)-N-(2-furylmethyl)amine
Supplier: Matrix Scientific
053174-500MG
053174-2.5G
101902-584EA
364.62
USD
101902-584
101902-586
N-(5-Bromo-4-methyl-2-pyridinyl)-N-(2-furylmethyl)amine
N-(5-Bromo-4-methyl-2-pyridinyl)-N-(2-furylmethyl)amine
Hazard: Irritant
Formula:
C₁₁H₁₁BrN₂O |
MDL Number:
MFCD13562615 CAS Number: 1220016-84-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...