4-[(2-Furylmethyl)amino]-3-nitrobenzoic acid
Supplier: Matrix Scientific
053173-500MG
053173-2.5G
101902-580EA
364.62
USD
101902-580
101902-582
4-[(2-Furylmethyl)amino]-3-nitrobenzoic acid
4-[(2-Furylmethyl)amino]-3-nitrobenzoic acid
Hazard: Irritant
Formula:
C₁₂H₁₀N₂O₅ |
MDL Number:
MFCD03982620 CAS Number: 284672-98-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...