Ethyl-3-amino-4-[(2-furylmethyl)amino]benzoate

Supplier: Matrix Scientific

053172-500MG 053172-2.5G
101902-576EA 364.62 USD
101902-576 101902-578
Ethyl-3-amino-4-[(2-furylmethyl)amino]benzoate
Ethyl-3-amino-4-[(2-furylmethyl)amino]benzoate

Hazard: Irritant

Formula: C₁₄H₁₆N₂O₃
MDL Number: MFCD13343609
CAS Number: 1220035-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR