3-Amino-4-chloro-N-phenylbenzenesulfonamide

Supplier: Matrix Scientific

053171-500MG 053171-2.5G
101902-572EA 364.62 USD
101902-572 101902-574
3-Amino-4-chloro-N-phenylbenzenesulfonamide
3-Amino-4-chloro-N-phenylbenzenesulfonamide

Hazard: Irritant

Formula: C₁₂H₁₁ClN₂O₂S
MDL Number: MFCD01911903
CAS Number: 94160-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR