N-Phenyl-4-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

053169-500MG 053169-2.5G
101902-564EA 384.62 USD
101902-564 101902-566
N-Phenyl-4-piperidinecarboxamide hydrochloride
N-Phenyl-4-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O
MDL Number: MFCD07324911
CAS Number: 73415-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR