N-Phenyl-3-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

053168-500MG 053168-2.5G
101902-560EA 364.62 USD
101902-560 101902-562
N-Phenyl-3-piperidinecarboxamide hydrochloride
N-Phenyl-3-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O
MDL Number: MFCD08246137
CAS Number: 173987-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR