N-Phenyl-2-piperidinecarboxamide hydrochloride
Supplier: Matrix Scientific
053167-500MG
053167-2.5G
101902-556EA
364.62
USD
101902-556
101902-558
N-Phenyl-2-piperidinecarboxamide hydrochloride
N-Phenyl-2-piperidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₇ClN₂O MW: 240.74 g/mol |
MDL Number:
MFCD13562613 CAS Number: 1246172-62-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...