N-Phenyl-2-pyrrolidinecarboxamide hydrochloride
Supplier: Matrix Scientific
053166-500MG
053166-2.5G
101902-552EA
364.62
USD
101902-552
101902-554
N-Phenyl-2-pyrrolidinecarboxamide hydrochloride
N-Phenyl-2-pyrrolidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₅ClN₂O |
MDL Number:
MFCD09863396 CAS Number: 1078162-90-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...