2-Amino-2-methyl-N-phenylpropanamide hydrochloride
Supplier: Matrix Scientific
053164-500MG
053164-2.5G
101902-544EA
364.62
USD
101902-544
101902-546
2-Amino-2-methyl-N-phenylpropanamide hydrochloride
2-Amino-2-methyl-N-phenylpropanamide hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₅ClN₂O |
MDL Number:
MFCD13562611 CAS Number: 1219964-09-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...