2-Amino-3-methyl-N-phenylbutanamide hydrochloride
Supplier: Matrix Scientific
053163-500MG
053163-2.5G
101902-540EA
364.62
USD
101902-540
101902-542
2-Amino-3-methyl-N-phenylbutanamide hydrochloride
2-Amino-3-methyl-N-phenylbutanamide hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₇ClN₂O MW: 228.72 g/mol |
MDL Number:
MFCD13562610 CAS Number: 635682-91-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...