3-Amino-N-phenylpropanamide hydrochloride

Supplier: Matrix Scientific

053162-500MG 053162-2.5G
101902-536EA 364.62 USD
101902-536 101902-538
3-Amino-N-phenylpropanamide hydrochloride
3-Amino-N-phenylpropanamide hydrochloride

Hazard: Irritant

Formula: C₉H₁₃ClN₂O
MDL Number: MFCD13193981
CAS Number: 115022-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR