2-(Methylamino)-N-phenylacetamide hydrochloride

Supplier: Matrix Scientific

053161-500MG 053161-2.5G
101902-532EA 364.62 USD
101902-532 101902-534
2-(Methylamino)-N-phenylacetamide hydrochloride
2-(Methylamino)-N-phenylacetamide hydrochloride

Hazard: Irritant

Formula: C₉H₁₃ClN₂O
MDL Number: MFCD10686932
CAS Number: 60565-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR