2-Amino-N-phenylacetamide hydrochloride
Supplier: Matrix Scientific
053159-500MG
053159-2.5G
101902-524EA
166.15
USD
101902-524
101902-526
2-Amino-N-phenylacetamide hydrochloride
2-Amino-N-phenylacetamide hydrochloride
Hazard: Irritant
Formula:
C₈H₁₁ClN₂O |
MDL Number:
MFCD00035439 CAS Number: 4801-39-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...