6-Chloro-N-phenyl-2-pyridinamine

Supplier: Matrix Scientific

053158-500MG 053158-2.5G
101902-520EA 364.62 USD
101902-520 101902-522
6-Chloro-N-phenyl-2-pyridinamine
6-Chloro-N-phenyl-2-pyridinamine

Hazard: Irritant

Formula: C₁₁H₉ClN₂
MDL Number: MFCD13562608
CAS Number: 854889-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR