5-Bromo-2-(phenylamino)pyridine

Supplier: Matrix Scientific

053157-500MG 053157-2.5G
101902-516EA 364.62 USD
101902-516 101902-518
5-Bromo-2-(phenylamino)pyridine
5-Bromo-2-(phenylamino)pyridine

Hazard: Irritant

Formula: C₁₁H₉BrN₂
MDL Number: MFCD12546457
CAS Number: 54904-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR