N-(5-Bromo-3-methyl-2-pyridinyl)-N-phenylamine

Supplier: Matrix Scientific

053156-500MG 053156-2.5G
101902-512EA 364.62 USD
101902-512 101902-514
N-(5-Bromo-3-methyl-2-pyridinyl)-N-phenylamine
N-(5-Bromo-3-methyl-2-pyridinyl)-N-phenylamine

Hazard: Irritant

Formula: C₁₂H₁₁BrN₂
MDL Number: MFCD13562607
CAS Number: 1219957-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR