2-Chloro-N-phenylnicotinamide
Supplier: Matrix Scientific
053155-500MG
053155-2.5G
101902-508EA
364.62
USD
101902-508
101902-510
2-Chloro-N-phenylnicotinamide
2-Chloro-N-phenylnicotinamide
Hazard: Irritant
Formula:
C₁₂H₉ClN₂O Melting Pt: 125 °C |
MDL Number:
MFCD00663736 CAS Number: 56149-29-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...