N2-Phenyl-2,3-pyridinediamine

Supplier: Matrix Scientific

053154-500MG 053154-2.5G
101902-504EA 364.62 USD
101902-504 101902-506
N2-Phenyl-2,3-pyridinediamine
N2-Phenyl-2,3-pyridinediamine

Hazard: Irritant

Formula: C₁₁H₁₁N₃
MDL Number: MFCD00823189
CAS Number: 41010-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR