2-Anilinonicotinic acid

Supplier: Matrix Scientific

Synonyms: 2-(Phenylamino)nicotinic acid

053153-500MG 053153-2.5G
101902-500EA 364.62 USD
101902-500 101902-502
2-Anilinonicotinic acid
2-Anilinonicotinic acid

Hazard: Irritant

Formula: C₁₂H₁₀N₂O₂
MW: 214.23 g/mol
MDL Number: MFCD00128791
CAS Number: 16344-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR