6-Chloro-N-phenyl-2-pyrazinamine

Supplier: Matrix Scientific

053151-500MG 053151-2.5G
101902-492EA 384.62 USD
101902-492 101902-494
6-Chloro-N-phenyl-2-pyrazinamine
6-Chloro-N-phenyl-2-pyrazinamine

Hazard: Irritant

Formula: C₁₀H₈ClN₃
MDL Number: MFCD13562606
CAS Number: 642459-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR