6-Chloro-N-phenyl-4-pyrimidinamine
Supplier: Matrix Scientific
053149-500MG
053149-2.5G
101902-484EA
384.62
USD
101902-484
101902-486
6-Chloro-N-phenyl-4-pyrimidinamine
6-Chloro-N-phenyl-4-pyrimidinamine
Hazard: Irritant
Formula:
C₁₀H₈ClN₃ |
MDL Number:
MFCD03861158 CAS Number: 69591-19-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...