N-(5-Bromo-4-methyl-2-pyridinyl)-N-phenylamine
Supplier: Matrix Scientific
053148-500MG
053148-2.5G
101902-480EA
364.62
USD
101902-480
101902-482
N-(5-Bromo-4-methyl-2-pyridinyl)-N-phenylamine
N-(5-Bromo-4-methyl-2-pyridinyl)-N-phenylamine
Hazard: Irritant
Formula:
C₁₂H₁₁BrN₂ |
MDL Number:
MFCD13562604 CAS Number: 1219957-62-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...