Ethyl-3-amino-4-anilinobenzoate

Supplier: Matrix Scientific

053146-500MG 053146-2.5G
101902-472EA 364.62 USD
101902-472 101902-474
Ethyl-3-amino-4-anilinobenzoate
Ethyl-3-amino-4-anilinobenzoate

Hazard: Irritant

Formula: C₁₅H₁₆N₂O₂
MDL Number: MFCD01156574
CAS Number: 68765-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR