3-Amino-4-chloro-N-(3-methoxypropyl)benzenesulfonamide
Supplier: Matrix Scientific
053145-500MG
053145-2.5G
101902-468EA
364.62
USD
101902-468
101902-470
3-Amino-4-chloro-N-(3-methoxypropyl)benzenesulfonamide
3-Amino-4-chloro-N-(3-methoxypropyl)benzenesulfonamide
Hazard: Irritant
Formula:
C₁₀H₁₅ClN₂O₃S MW: 278.76 g/mol |
MDL Number:
MFCD11206522 CAS Number: 1094269-51-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...