N-(3-Methoxypropyl)-4-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

053143-500MG 053143-2.5G
101902-460EA 364.62 USD
101902-460 101902-462
N-(3-Methoxypropyl)-4-piperidinecarboxamide hydrochloride
N-(3-Methoxypropyl)-4-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₁ClN₂O₂
MW: 236.74 g/mol
MDL Number: MFCD12912987
CAS Number: 1193389-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR