N-(3-Methoxypropyl)-2-piperidinecarboxamide hydrochloride
Supplier: Matrix Scientific
053141-500MG
053141-2.5G
101902-452EA
364.62
USD
101902-452
101902-454
N-(3-Methoxypropyl)-2-piperidinecarboxamide hydrochloride
N-(3-Methoxypropyl)-2-piperidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₀H₂₁ClN₂O₂ MW: 236.74 g/mol |
MDL Number:
MFCD13562601 CAS Number: 1236256-96-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...