N-(3-Methoxypropyl)-2-pyrrolidinecarboxamide hydrochloride

Supplier: Matrix Scientific

053140-500MG 053140-2.5G
101902-448EA 364.62 USD
101902-448 101902-450
N-(3-Methoxypropyl)-2-pyrrolidinecarboxamide hydrochloride
N-(3-Methoxypropyl)-2-pyrrolidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₉H₁₉ClN₂O₂
MDL Number: MFCD13562600
CAS Number: 1236261-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR