2-Amino-N-(3-methoxypropyl)-3-phenylpropanamide hydrochloride

Supplier: Matrix Scientific

053139-500MG 053139-2.5G
101902-444EA 364.62 USD
101902-444 101902-446
2-Amino-N-(3-methoxypropyl)-3-phenylpropanamide hydrochloride
2-Amino-N-(3-methoxypropyl)-3-phenylpropanamide hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₁ClN₂O₂
MDL Number: MFCD13562599
CAS Number: 1236256-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR