2-Amino-N-(3-methoxypropyl)propanamide hydrochloride

Supplier: Matrix Scientific

053134-500MG 053134-2.5G
101902-424EA 364.62 USD
101902-424 101902-426
2-Amino-N-(3-methoxypropyl)propanamide hydrochloride
2-Amino-N-(3-methoxypropyl)propanamide hydrochloride

Hazard: Irritant

Formula: C₇H₁₇ClN₂O₂
MDL Number: MFCD13562594
CAS Number: 1236267-66-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR