6-Chloro-N-(3-methoxypropyl)-2-pyridinamine

Supplier: Matrix Scientific

053132-500MG 053132-2.5G
101902-416EA 364.62 USD
101902-416 101902-418
6-Chloro-N-(3-methoxypropyl)-2-pyridinamine
6-Chloro-N-(3-methoxypropyl)-2-pyridinamine

Hazard: Irritant

Formula: C₉H₁₃ClN₂O
MDL Number: MFCD13562592
CAS Number: 29449-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR