5-Bromo-N-(3-methoxypropyl)-2-pyridinamine

Supplier: Matrix Scientific

053131-500MG 053131-2.5G
101902-412EA 364.62 USD
101902-412 101902-414
5-Bromo-N-(3-methoxypropyl)-2-pyridinamine
5-Bromo-N-(3-methoxypropyl)-2-pyridinamine

Hazard: Irritant

Formula: C₉H₁₃BrN₂O
MDL Number: MFCD11206566
CAS Number: 1094433-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR