2-Chloro-N-(3-methoxypropyl)nicotinamide
Supplier: Matrix Scientific
053129-500MG
053129-2.5G
101902-404EA
364.62
USD
101902-404
101902-406
2-Chloro-N-(3-methoxypropyl)nicotinamide
2-Chloro-N-(3-methoxypropyl)nicotinamide
Hazard: Irritant
Formula:
C₁₀H₁₃ClN₂O₂ |
MDL Number:
MFCD03398037 CAS Number: 547706-96-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...