2-[(3-Methoxypropyl)amino]nicotinic acid

Supplier: Matrix Scientific

053127-500MG 053127-2.5G
101902-396EA 364.62 USD
101902-396 101902-398
2-[(3-Methoxypropyl)amino]nicotinic acid
2-[(3-Methoxypropyl)amino]nicotinic acid

Hazard: Irritant

Formula: C₁₀H₁₄N₂O₃
MDL Number: MFCD11104055
CAS Number: 1152596-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR