2-Chloro-6-((3-methoxypropyl)amino)pyrazine

Supplier: Matrix Scientific

053125-500MG 053125-2.5G
101902-388EA 364.62 USD
101902-388 101902-390
2-Chloro-6-((3-methoxypropyl)amino)pyrazine
2-Chloro-6-((3-methoxypropyl)amino)pyrazine

Hazard: Irritant

Formula: C₈H₁₂ClN₃O
MDL Number: MFCD09800963
CAS Number: 941294-49-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR