3-Chloro-N-(3-methoxypropyl)-5-(trifluoromethyl)-2-pyridinamine

Supplier: Matrix Scientific

053124-500MG 053124-2.5G
101902-384EA 364.62 USD
101902-384 101902-386
3-Chloro-N-(3-methoxypropyl)-5-(trifluoromethyl)-2-pyridinamine
3-Chloro-N-(3-methoxypropyl)-5-(trifluoromethyl)-2-pyridinamine

Hazard: Irritant

Formula: C₁₀H₁₂ClF₃N₂O
MDL Number: MFCD00172036
CAS Number: 338406-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR