6-Chloro-N-(3-methoxypropyl)-4-pyrimidinamine

Supplier: Matrix Scientific

Synonyms: 6-Chloro-N-(3-methoxypropyl)pyrimidin-4-amine

053123-500MG 053123-2.5G
101902-380EA 364.62 USD
101902-380 101902-382
6-Chloro-N-(3-methoxypropyl)-4-pyrimidinamine
6-Chloro-N-(3-methoxypropyl)-4-pyrimidinamine

Hazard: Irritant

Formula: C₈H₁₂ClN₃O
MW: 201.66 g/mol
MDL Number: MFCD09800960
CAS Number: 941294-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR