5-Bromo-N-(3-methoxypropyl)-4-methyl-2-pyridinamine

Supplier: Matrix Scientific

053122-500MG 053122-2.5G
101902-376EA 364.62 USD
101902-376 101902-378
5-Bromo-N-(3-methoxypropyl)-4-methyl-2-pyridinamine
5-Bromo-N-(3-methoxypropyl)-4-methyl-2-pyridinamine

Hazard: Irritant

Formula: C₁₀H₁₅BrN₂O
MDL Number: MFCD13562590
CAS Number: 1220028-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR