4-[(3-Methoxypropyl)amino]-3-nitrobenzoic acid

Supplier: Matrix Scientific

053121-500MG 053121-2.5G
101902-372EA 364.62 USD
101902-372 101902-374
4-[(3-Methoxypropyl)amino]-3-nitrobenzoic acid
4-[(3-Methoxypropyl)amino]-3-nitrobenzoic acid

Hazard: Irritant

Formula: C₁₁H₁₄N₂O₅
MDL Number: MFCD11543601
CAS Number: 1051102-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR