Ethyl-3-amino-4-[(3-methoxypropyl)amino]benzoate

Supplier: Matrix Scientific

053120-500MG 053120-2.5G
101902-368EA 364.62 USD
101902-368 101902-370
Ethyl-3-amino-4-[(3-methoxypropyl)amino]benzoate
Ethyl-3-amino-4-[(3-methoxypropyl)amino]benzoate

Hazard: Irritant

Formula: C₁₃H₂₀N₂O₃
MDL Number: MFCD13344581
CAS Number: 1220038-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR