N-(2-Hydroxy-1,1-dimethylethyl)-4,5,6,7-tetra-hydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide HCl
Supplier: Matrix Scientific
053118-500MG
053118-2.5G
101902-360EA
384.62
USD
101902-360
101902-362
N-(2-Hydroxy-1,1-dimethylethyl)-4,5,6,7-tetra-hydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide HCl
N-(2-Hydroxy-1,1-dimethylethyl)-4,5,6,7-tetra-hydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide HCl
Hazard: Irritant
Formula:
C₁₁H₁₉ClN₄O₂ |
MDL Number:
MFCD13562589 CAS Number: 1220030-25-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...