N-(2-Hydroxy-1,1-dimethylethyl)-4-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

053117-500MG 053117-2.5G
101902-356EA 364.62 USD
101902-356 101902-358
N-(2-Hydroxy-1,1-dimethylethyl)-4-piperidinecarboxamide hydrochloride
N-(2-Hydroxy-1,1-dimethylethyl)-4-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₁ClN₂O₂
MDL Number: MFCD08457445
CAS Number: 40051-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR