N-(2-Hydroxy-1,1-dimethylethyl)-3-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

053116-500MG 053116-2.5G
101902-352EA 364.62 USD
101902-352 101902-354
N-(2-Hydroxy-1,1-dimethylethyl)-3-piperidinecarboxamide hydrochloride
N-(2-Hydroxy-1,1-dimethylethyl)-3-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₁ClN₂O₂
MDL Number: MFCD13562588
CAS Number: 1220037-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR