N-(2-Hydroxy-1,1-dimethylethyl)-2-piperidinecarboxamide hydrochloride
Supplier: Matrix Scientific
053115-500MG
053115-2.5G
101902-348EA
364.62
USD
101902-348
101902-350
N-(2-Hydroxy-1,1-dimethylethyl)-2-piperidinecarboxamide hydrochloride
N-(2-Hydroxy-1,1-dimethylethyl)-2-piperidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₀H₂₁ClN₂O₂ |
MDL Number:
MFCD13562587 CAS Number: 1236255-13-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...