N-(2-Hydroxy-1,1-dimethylethyl)-2-pyrrolidinecarboxamide hydrochloride
Supplier: Matrix Scientific
053114-500MG
053114-2.5G
101902-344EA
364.62
USD
101902-344
101902-346
N-(2-Hydroxy-1,1-dimethylethyl)-2-pyrrolidinecarboxamide hydrochloride
N-(2-Hydroxy-1,1-dimethylethyl)-2-pyrrolidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₉H₁₉ClN₂O₂ |
MDL Number:
MFCD13562586 CAS Number: 1236266-84-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...